SY3H3D67DB00 Synthonix Corporation

2-(3-Ethoxy-4-(ethoxycarbonyl)phenyl)acetic acid

  • MDL number MFCD08704233
  • Cas Number : 99469-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56996.html

Safety Information

signalword Warning
MSDS https://synthonix.com/56996.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart