SY3H3D67DB16 Synthonix Corporation

4-Ethyl-1,1'-Biphenyl

  • MDL number MFCD00010236
  • Cas Number : 5707-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Lump / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57632.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57632.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart