SY3H3D67DB1A Synthonix Corporation

Ethyl 4-ethoxyquinoline-7-carboxylate

  • Cas Number : 1688656-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57679.html

Safety Information

MSDS https://synthonix.com/57679.html
Availability details for
Quantity
  • Availability
  • Add to Cart