SY3H3D67DB1B Synthonix Corporation

(4'-Ethyl-[1,1'-biphenyl]-4-yl)boronic acid

  • MDL number MFCD04038068
  • Cas Number : 153035-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57690.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57690.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart