SY3H3D67DB1F Synthonix Corporation

Ethyl 6-methyl-2-oxo-4-phenyl-1,2,3,4-tetrahydropyrimidine-5-carboxylate

  • Cas Number : 5395-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57709.html

Safety Information

MSDS https://synthonix.com/57709.html
Availability details for
Quantity
  • Availability
  • Add to Cart