SY3H3D67DB21 Synthonix Corporation

4-(4-Ethoxyphenyl)cyclohexanone

  • Cas Number : 163671-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57806.html

Safety Information

MSDS https://synthonix.com/57806.html
Availability details for
Quantity
  • Availability
  • Add to Cart