SY3H3D67DB22 Synthonix Corporation

Ethyl 2-Acetyl-4-phenylbutanoate

  • Cas Number : 5337-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57811.html

Safety Information

MSDS https://synthonix.com/57811.html
Availability details for
Quantity
  • Availability
  • Add to Cart