SY3H3D67DB23 Synthonix Corporation

Ethyl isobutyl phthalate

  • Cas Number : 94491-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57816.html

Safety Information

MSDS https://synthonix.com/57816.html
Availability details for
Quantity
  • Availability
  • Add to Cart