SY3H3D67DBC7 Synthonix Corporation

Ethyl 2,4-dihydroxy-6-pentylbenzoate

  • Cas Number : 38862-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57912.html

Safety Information

MSDS https://synthonix.com/57912.html
Availability details for
Quantity
  • Availability
  • Add to Cart