SY3H3D67DBDB Synthonix Corporation

Ethyl 2-(7-methoxynaphthalen-1-yl)acetate

  • MDL number MFCD12756805
  • Cas Number : 6836-21-1
  • Molecular Weight : 244.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (7-methoxy-1-naphthyl)acetate
Inchi Code 1S/C15H16O3/c1-3-18-15(16)9-12-6-4-5-11-7-8-13(17-2)10-14(11)12/h4-8,10H,3,9H2,1-2H3
Inchi Key VPXDDUIYFMKQBI-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Colorless or White to yellow powder or crystals or liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/58282.html

Safety Information

signalword Warning
pcodes P280,P305,P351,P338
MSDS https://synthonix.com/58282.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart