SY3H3D67DBE1 Synthonix Corporation

Ethyl 1-benzylpiperidine-3-carboxylate

  • MDL number MFCD00191221
  • Cas Number : 72551-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58395.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58395.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart