SY3H3D67DBE2 Synthonix Corporation

(S)-Ethyl 2-(4-benzylmorpholin-3-yl)acetate

  • MDL number MFCD28963475
  • Cas Number : 1799439-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58398.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58398.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart