SY3H3D67DBE4 Synthonix Corporation

Ethyl 2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)acetate

  • MDL number MFCD00185560
  • Cas Number : 14028-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58413.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58413.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart