SY3H3D67DBF7 Synthonix Corporation

Ethyl 2-(diphenoxyphosphoryl)acetate

  • MDL number MFCD01321165
  • Cas Number : 16139-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Clear liquid / Colour: Very pale yellow - Yellow - Brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58701.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58701.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart