SY3H3D67DBFC Synthonix Corporation

Ethyl N-Cbz-piperidine-4-carboxylate

  • Cas Number : 160809-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58758.html

Safety Information

MSDS https://synthonix.com/58758.html
Availability details for
Quantity
  • Availability
  • Add to Cart