SY3H3D67DC06 Synthonix Corporation

4-Ethynyl-4'-propyl-1,1'-biphenyl

  • MDL number MFCD18072535
  • Cas Number : 360768-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58925.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58925.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart