SY3H3D67DC08 Synthonix Corporation

1-Ethoxy-4-(p-tolylethynyl)benzene

  • MDL number MFCD01817758
  • Cas Number : 116903-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58937.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58937.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart