SY3H3D67DC5D Synthonix Corporation

3-Ethyl-2-(2-(phenylamino)vinyl)benzo[d]thiazol-3-ium iodide

  • MDL number MFCD01232746
  • Cas Number : 60126-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59111.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59111.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart