SY3H3D67DC68 Synthonix Corporation

(4-ethoxyphenyl) 4-propylcyclohexane-1-carboxylate

  • MDL number MFCD22422984
  • Cas Number : 67589-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59307.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59307.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart