SY3H3D67DC69 Synthonix Corporation

Ethyl 4-(5-(bis(2-hydroxyethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoate

  • Cas Number : 3543-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59312.html

Safety Information

MSDS https://synthonix.com/59312.html
Availability details for
Quantity
  • Availability
  • Add to Cart