SY3H3D67DC6D Synthonix Corporation

Ethyl 6-bromo-5-hydroxy-1-methyl-2-((phenylthio)methyl)-1H-Indole-3-carboxylate

  • MDL number MFCD00378136
  • Cas Number : 131707-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59397.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59397.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart