SY3H3D67DC6F Synthonix Corporation

Ethyl 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylate

  • Cas Number : 105404-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/59418.html

Safety Information

MSDS https://synthonix.com/59418.html
Availability details for
Quantity
  • Availability
  • Add to Cart