SY3H3D67DC73 Synthonix Corporation

4-Ethoxyphenyl 4-((butoxycarbonyl)oxy)benzoate

  • MDL number MFCD00051359
  • Cas Number : 16494-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/59553.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/59553.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart