SY3H3D67DC8A Synthonix Corporation

4-Fluoro-1-iodo-2-methylbenzene

  • MDL number MFCD00039397
  • Cas Number : 66256-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Colorless to pale-red to yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39376.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39376.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart