SY3H3D67DC9B Synthonix Corporation

3-(4-Fluorophenyl)-1-isopropyl-1H-indole

  • MDL number MFCD01075733
  • Cas Number : 93957-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: Very pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39740.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39740.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart