SY3H3D67DCA4 Synthonix Corporation

2-Fluoro-3,4-diiodopyridine

  • Cas Number : 153034-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40541.html

Safety Information

MSDS https://synthonix.com/40541.html
Availability details for
Quantity
  • Availability
  • Add to Cart