SY3H3D67DCF4 Synthonix Corporation

3-Fluoro-2-nitropyridine

  • MDL number MFCD04114127
  • Cas Number : 54231-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40585.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40585.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart