SY3H3D67DCF5 Synthonix Corporation

3-Fluoro-5-nitropyridine

  • Cas Number : 1060804-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40586.html

Safety Information

MSDS https://synthonix.com/40586.html
Availability details for
Quantity
  • Availability
  • Add to Cart