SY3H3D67DCFF Synthonix Corporation

4-Fluoro-5-iodopyridin-2-amine

  • Cas Number : 1708974-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40752.html

Safety Information

MSDS https://synthonix.com/40752.html
Availability details for
Quantity
  • Availability
  • Add to Cart