SY3H3D67DD0A Synthonix Corporation

(5-Formylfuran-3-yl)boronic acid

  • MDL number MFCD01319004
  • Cas Number : 62306-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41013.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41013.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart