SY3H3D67DD0B Synthonix Corporation

3-Fluoro-2-hydrazinylpyridine

  • MDL number MFCD09998071
  • Cas Number : 887266-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to off-white to brown powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41082.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41082.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart