SY3H3D67DD0D Synthonix Corporation

6-Fluoro-5-methylpyrimidin-4-amine

  • MDL number MFCD10000602
  • Cas Number : 18260-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41084.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41084.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart