SY3H3D67DD1D Synthonix Corporation

4-Fluoro-2-iodo-1-nitrobenzene

  • MDL number MFCD16658618
  • Cas Number : 41860-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41957.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41957.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart