SY3H3D67DD24 Synthonix Corporation

3-Fluoro-5-(trifluoromethyl)pyridin-2-amine

  • Cas Number : 852062-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42082.html

Safety Information

MSDS https://synthonix.com/42082.html
Availability details for
Quantity
  • Availability
  • Add to Cart