SY3H3D67DD38 Synthonix Corporation

3-Fluoro-4-iodophenol

  • MDL number MFCD12407087
  • Cas Number : 122927-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown to Orange Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42369.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42369.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart