SY3H3D67DD43 Synthonix Corporation

5-Fluoro-1-methyl-3-nitropyridin-2(1H)-one

  • MDL number MFCD28099326
  • Cas Number : 1616526-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42443.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42443.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart