SY3H3D67DD4F Synthonix Corporation

3-Fluoro-2-iodoaniline hydrochloride

  • Cas Number : 122455-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42597.html

Safety Information

MSDS https://synthonix.com/42597.html
Availability details for
Quantity
  • Availability
  • Add to Cart