SY3H3D67DD54 Synthonix Corporation

2-Fluorobenzene-1,3-diol

  • Cas Number : 103068-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42703.html

Safety Information

MSDS https://synthonix.com/42703.html
Availability details for
Quantity
  • Availability
  • Add to Cart