SY3H3D67DD57 Synthonix Corporation

5-Fluoropicolinohydrazide

  • Cas Number : 1254073-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/42828.html

Safety Information

MSDS https://synthonix.com/42828.html
Availability details for
Quantity
  • Availability
  • Add to Cart