SY3H3D67DD62 Synthonix Corporation

2-Fluoro-3-(hydroxymethyl)pyridine

  • MDL number MFCD03092920
  • Cas Number : 131747-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/42930.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/42930.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart