SY3H3D67DD70 Synthonix Corporation

3-Fluoro-5-methylpyridin-2-amine

  • MDL number No MDL found
  • Cas Number : 1211590-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43015.html

Safety Information

MSDS https://synthonix.com/43015.html
Availability details for
Quantity
  • Availability
  • Add to Cart