SY3H3D67DD77 Synthonix Corporation

6-Fluoro-3-methoxypyridin-2-amine

  • MDL number MFCD28722398
  • Cas Number : 1805602-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/43084.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/43084.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart