SY3H3D67DD7A Synthonix Corporation

5-Fluoro-2,6-dimethylpyrimidin-4-amine

  • Cas Number : 1472019-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/43284.html

Safety Information

MSDS https://synthonix.com/43284.html
Availability details for
Quantity
  • Availability
  • Add to Cart