SY3H3D67DD83 Synthonix Corporation

1-Fluoro-4-iodo-2-(trifluoromethyl)benzene

  • MDL number MFCD00236678
  • Cas Number : 59382-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - yellow - pale-green - black
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44045.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44045.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart