SY3H3D67DD87 Synthonix Corporation

3-Fluoro-4-nitrobenzonitrile

  • MDL number MFCD04973417
  • Cas Number : 218632-01-0
  • Molecular Weight : 166.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-4-nitrobenzonitrile
Inchi Code 1S/C7H3FN2O2/c8-6-3-5(4-9)1-2-7(6)10(11)12/h1-3H
Inchi Key OZXCOGPRBUSXLE-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/44071.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44071.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart