SY3H3D67E33F Synthonix Corporation

5-Fluorobenzo[d]thiazole

  • Cas Number : 1644-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/44438.html

Safety Information

MSDS https://synthonix.com/44438.html
Availability details for
Quantity
  • Availability
  • Add to Cart