SY3H3D67E340 Synthonix Corporation

5-Fluorobenzo[d]thiazole-2-thiol

  • MDL number MFCD02181177
  • Cas Number : 155559-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form White to yellow to pale-brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44439.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44439.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart