SY3H3D67E341 Synthonix Corporation

6-Fluorobenzo[d]thiazole-2(3H)-thione

  • MDL number MFCD08061254
  • Cas Number : 80087-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44440.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44440.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart