SY3H3D67E344 Synthonix Corporation

6-Fluorobenzo[d]isoxazol-3(2H)-one

  • MDL number MFCD13180537
  • Cas Number : 99822-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44455.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44455.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart