SY3H3D67E34D Synthonix Corporation

2-Fluoro-4-nitrobenzoic acid

  • MDL number MFCD00275565
  • Cas Number : 403-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44508.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44508.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart